Order Entry
Austria
Orders LinkContactUsLinkComponent
Buckets

Buckets

Transparent, 15 L, 340 mm
Catalog # BRND72378
Supplier:   Brand
Buckets
Catalog # BRND72378
Supplier:  Brand
Buckets
Buckets

Transparent, 15 L, 340 mm

Price
38,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP, translucent.

  • Graduated
  • Very good thermal resistance
  • Very good chemical resistance

Universal multipurpose bucket with spout, reinforced rim and handle.

Specifications
  • Capacity:
    15 L
  • Colour:
    Transparent
  • Height:
    340 mm