Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Celite® 545, filter aid

Plastic bottle, 500 g
Catalog # 349675000.
Health
Celite® 545, filter aid
Catalog # 349675000.
CAS Number:  68855-54-9
Celite® 545, filter aid
Celite® 545, filter aid

Plastic bottle, 500 g

CAS Number:  68855-54-9
Price
47,40 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  DE, Diatomaceous earth, flux-calcined
  • CAS Number:68855-54-9
  • Formula:SiO₂
  • MDL Number:MFCD00132803
  • Molecular Weight:60.08 g/mol
  • Storage Temperature:Ambient
  • EINECS:272-489-0
Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Particle size:
    ≤25 % (Retained on 150 mesh)
  • Wet density:
    255 to 370 g/l
  • pH:
    9.0 to 11.0 (10 % slurry)
  • Permeability:
    2.6 to 6.5 Darcy
  • Calcium oxide (CaO):
    0.3 % (Typical)
  • Magnesium oxide (MgO):
    0.3 % (Typical)
  • Silicon dioxide (SiO2):
    91.5 % (Typical)
  • Aluminium oxide (Al2O3):
    1.0 % (Typical)
  • Iron(III)oxide (Fe2O3):
    1.5 % (Typical)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications