Order Entry
Austria
Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes

Sterilisation Boxes for Pipettes

Square base, 70×70×282 mm
Catalog # 612-1627
Supplier:   BOCHEM
Sterilisation Boxes for Pipettes
Catalog # 612-1627
Supplier:  BOCHEM
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes

Square base, 70×70×282 mm

Price
113,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, with square base, autoclavable.

  • Can be autoclaved at 121 °C and sterilised at 205 °C

Bottom and underside of the lid are coated in silicone to minimise the risk of damaging the pipettes during transport and sterilisation

Specifications
  • Description:
    Square base
  • W×D×H:
    70×70×282 mm