Retort rods, without thread
12 mm, 500 mm
Catalog # BOCH5120
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10.
- Rods for stand bases
- Without thread
Specifications
- Ø:12 mm
- Length:500 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



