Order Entry
Austria
Orders LinkContactUsLinkComponent
Stand bases

Stand bases

2,4 kg, 200×315×3 mm
Catalog # BOCH5003
Supplier:   BOCHEM
Stand bases
Catalog # BOCH5003
Supplier:  BOCHEM
Stand bases
Stand bases

2,4 kg, 200×315×3 mm

Price
143,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stand bases made of stainless steel 18/10 with expanded, flat and heavy feet for stability.

  • With M 10 threaded hole (off-centre)
  • One adjustable foot
  • Anti-slip rubber protection

Certifications: DIN 12892

Specifications
  • W×D×H:
    200×315×3 mm
  • Weight:
    2,4 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.