Orders LinkContactUsLinkComponent
Microscopy scissors
Microscopy scissors
Curved, sharp/sharp, 100 mm
Catalog # 233-2123
Supplier:   BOCHEM
Microscopy scissors
Catalog # 233-2123
Supplier:  BOCHEM
Microscopy scissors
Microscopy scissors
Curved, sharp/sharp, 100 mm
Price
5,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Corrosion resistant
  • Autoclavable
  • Very sharp tips
Specifications
  • Type:
    Curved, sharp/sharp
  • Length:
    100 mm