Orders LinkContactUsLinkComponent
Laboratory scissors
Laboratory scissors
250 mm
Catalog # BOCH4023
Supplier:   BOCHEM
Laboratory scissors
Catalog # BOCH4023
Supplier:  BOCHEM
Laboratory scissors
Laboratory scissors
250 mm
Price
14,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, with blue plastic handles.
  • Corrosion resistant
  • Ergonomically shaped handle
Specifications
  • Length:
    250 mm