Order Entry
Austria
Orders LinkContactUsLinkComponent
Spoon spatulas

Spoon spatulas

180 mm, 25×8 mm
Catalog # 231-1051
Supplier:   BOCHEM
Spoon spatulas
Catalog # 231-1051
Supplier:  BOCHEM
Spoon spatulas
Spoon spatulas

180 mm, 25×8 mm

Price
2,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Universal spoon spatula designed for a wide range of uses.

  • One end is shaped like a spoon and the other end like a spatula
  • With rounded handle
  • High thermal resistance, autoclavable
  • Excellent resistance to chemicals
  • Quick and easy to clean

Documents

Specifications
  • Length:
    180 mm
  • Spoon size:
    25×8 mm