Orders LinkContactUsLinkComponent
Forceps with guide pin
Bent, sharp, 115 mm
Catalog # 232-0018
Supplier:   BOCHEM
Forceps with guide pin
Catalog # 232-0018
Supplier:  BOCHEM
Forceps with guide pin
Forceps with guide pin
Bent, sharp, 115 mm
Price
29,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, with PTFE coating.
  • Bent, with guide pin
  • Smooth, coated surface to prevent contamination and facilitate cleaning
Specifications
  • Type:
    Bent, sharp
  • Length:
    115 mm