Orders LinkContactUsLinkComponent
Forceps
Forceps
Blunt, 130 mm
Catalog # BOCH1801
Supplier:   BOCHEM
Forceps
Catalog # BOCH1801
Supplier:  BOCHEM
Forceps
Forceps
Blunt, 130 mm
Price
29,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very high chemical and thermal resistance
  • Smooth surface prevents any contamination and makes cleaning easier
Specifications
  • Type:
    Blunt
  • Length:
    130 mm