Forceps, precision
Precision forceps, smooth tips, stainless steel, Straight, extra sharp, 130 mm
Catalog # 232-0006
Supplier: BOCHEM
Jump to:
Product Details & Documents
These non serrated precision forceps are available in pure stainless steel or with PTFE coating.
Specifications
- Type:Straight, extra sharp
- Description:Precision forceps, smooth tips, stainless steel
- Length:130 mm