Orders LinkContactUsLinkComponent
Hoffmann pinch cocks
Hoffmann pinch cocks
38 mm
Catalog # 229-0069
Supplier:   BOCHEM
Hoffmann pinch cocks
Catalog # 229-0069
Supplier:  BOCHEM
Hoffmann pinch cocks
Hoffmann pinch cocks
38 mm
Price
7,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Nickel-plated brass, with flap.
Documents
Specifications
  • Seq:
    06
  • Bandwidth:
    38 mm