Order Entry
Austria
Orders LinkContactUsLinkComponent
Wire baskets, square and rectangular

Wire baskets, square and rectangular

100×150×100 mm
Catalog # BOCH10023
Supplier:   BOCHEM
Wire baskets, square and rectangular
Catalog # BOCH10023
Supplier:  BOCHEM
Wire baskets, square and rectangular
Wire baskets, square and rectangular

100×150×100 mm

Price
51,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.

  • Spot-welded grid 8×8×1 mm
  • Reinforced edges
  • Very good chemical and temperature resistance
Specifications
  • W×D×H:
    100×150×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.