Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
500 ml, 24/29 NS
Catalog # BIBB443054N
Supplier:   SciLabware
Separating funnels, Gilson
Catalog # BIBB443054N
Supplier:  SciLabware
Separating funnels, Gilson
Separating funnels, Gilson
500 ml, 24/29 NS
Price
146,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.
Specifications
  • Cone size:
    24/29 NS
  • Capacity:
    500 ml
  • Seq:
    11