Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Flasks, flat bottom, narrow neck

1000 ml, 200 mm, 36 mm, 131 mm
  214-0347
 :   SciLabware
Flasks, flat bottom, narrow neck
  214-0347
Flasks, flat bottom, narrow neck
Flasks, flat bottom, narrow neck

1000 ml, 200 mm, 36 mm, 131 mm

 
20,90 €
   
1 * 1 ST
 

PYREX®, borosilicate glass, clear.

  • High resistance to thermal shock
  • Excellent chemical durability for contamination-free performance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Narrow neck flasks with flat base for standing on benches and hotplates. Marked with capacity and a white labelling area.

 : ISO 1773

 
  • Ext.Ø:
    131 mm
  • Capacity:
    1000 ml
  • Height:
    200 mm
  • Seq:
    05
  • Neck I-Ø:
    36 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.