Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Rectangular cover glasses

Coverslips, 13×54 mm, 0,13 - 0,16 mm
  BELC1916-13054A
Marketsource
Rectangular cover glasses
  BELC1916-13054A
Marketsource
Rectangular cover glasses
Rectangular cover glasses

Coverslips, 13×54 mm, 0,13 - 0,16 mm

 
177,00 €
   
1 * 100 ST
 

These rectangular cover glasses are designed for use with culture tubes.

 
  • Description:
    Coverslips
  • L×W:
    13×54 mm
  • Seq:
    003
  • Thickness:
    0,13 - 0,16 mm