Orders LinkContactUsLinkComponent
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
250 ml grinding chamber, Use with double beater 431-2113. Polycarbonate with stainless steel inlet. Not suitable for Use with liquid nitrogen
Catalog # 412-1112
Supplier:   IKA
Accessories for analytical Mill, A 11 basic
Catalog # 412-1112
Supplier:  IKA
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
250 ml grinding chamber, Use with double beater 431-2113. Polycarbonate with stainless steel inlet. Not suitable for Use with liquid nitrogen
Price
738,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    250 ml grinding chamber
  • Seq:
    02
  • For:
    Use with double beater 431-2113. Polycarbonate with stainless steel inlet. Not suitable for Use with liquid nitrogen