Order Entry
Austria
Orders LinkContactUsLinkComponent
Volumetric flasks, class B

Volumetric flasks, class B

1000 ml, 24/29, 310 mm
Catalog # HIRS7560193
Supplier:   HIRSCHMANN
Volumetric flasks, class B
Catalog # HIRS7560193
Supplier:  HIRSCHMANN
Volumetric flasks, class B
Volumetric flasks, class B

1000 ml, 24/29, 310 mm

Price
27,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

PMP, translucent, with PP NS-stopper.

Specifications
  • NS:
    24/29
  • Capacity:
    1000 ml
  • Height:
    310 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.