Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Staining rack for 10 slides, glass, without handle, - mm
Catalog # 631-9321
Staining jars
Catalog # 631-9321
Staining jars
Staining jars
Staining rack for 10 slides, glass, without handle, - mm
Price
17,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Soda-lime glass.
  • For 10 standard slides  (76×26 mm)
Staining system contains staining jar, rack and handle made from stainless steel.
Ordering information: Insert and handle must be ordered separately.
Specifications
  • Description:
    Staining rack for 10 slides, glass, without handle
  • W×D×H:
    - mm