Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Staining jars

Staining dish with lid, 105×85×80 mm
  631-9328
Staining jars
  631-9328
Staining jars
Staining jars

Staining dish with lid, 105×85×80 mm

 
25,80 €
   
1 * 1 ST
 

Soda-lime glass.

  • For 10 standard slides  (76×26 mm)

Staining system contains staining jar, rack and handle made from stainless steel.

 : Insert and handle must be ordered separately.

 
  • Description:
    Staining dish with lid
  • W×D×H:
    105×85×80 mm