Staining racks
Staining rack, extendable, Plexiglas®
631-0336
Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or dishes of 380 to 620 mm in length.
- Description:Staining rack, extendable, Plexiglas®