Order Entry
Austria
Orders LinkContactUsLinkComponent
Cover glass forceps

Cover glass forceps

160 mm
Catalog # 232-2183
Supplier:   HAMMACHER KARL
Cover glass forceps
Catalog # 232-2183
Supplier:  HAMMACHER KARL
Cover glass forceps
Cover glass forceps

160 mm

Price
84,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).

  • For microscopic slides
  • Resistant to corrosion
  • Anti-magnetic
Specifications
  • Length:
    160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.