Orders LinkContactUsLinkComponent
Light forceps
Light forceps
Sharp, 100 mm
Catalog # 232-2119
Supplier:   HAMMACHER KARL
Light forceps
Catalog # 232-2119
Supplier:  HAMMACHER KARL
Light forceps
Light forceps
Sharp, 100 mm
Price
58,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Light forceps made of watch-spring sheet-steel.
Specifications
  • Type:
    Sharp
  • Length:
    100 mm
  • Seq:
    02