Orders LinkContactUsLinkComponent
Light forceps
Light forceps
Blunt, 100 mm
Catalog # 232-2120
Supplier:   HAMMACHER KARL
Light forceps
Catalog # 232-2120
Supplier:  HAMMACHER KARL
Light forceps
Light forceps
Blunt, 100 mm
Price
52,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Light forceps made of watch-spring sheet-steel.
Specifications
  • Type:
    Blunt
  • Length:
    100 mm
  • Seq:
    01