Orders LinkContactUsLinkComponent
Dissecting needles
Dissecting needles
Dissecting needle, Straight, fine, 130 mm
Catalog # HAMMHSO004-13
Supplier:   HAMMACHER KARL
Dissecting needles
Catalog # HAMMHSO004-13
Supplier:  HAMMACHER KARL
Dissecting needles
Dissecting needles
Dissecting needle, Straight, fine, 130 mm
Price
48,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
These dissecting needles are made of stainless steel.
Specifications
  • Form:
    Straight, fine
  • Description:
    Dissecting needle
  • Length:
    130 mm
  • Seq:
    01