Staining forceps
Bent, serrated, fine tips, 160 mm
Catalog # HAMML-HSC012-05
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Staining forceps made from stainless steel.
Specifications
- Type:Bent, serrated, fine tips
- Length:160 mm
- Seq:03