Universal scissors, straight
Sharp-blunt, 125 mm
Catalog # HAMMHSB952-12
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Stainless steel or chromium-plated steel.
- Sharp-blunt
- For a wide range of applications
- Corrosion resistant
- Easy action
Specifications
- Type:Sharp-blunt
- Length:125 mm
- Seq:01