Orders LinkContactUsLinkComponent
Iris scissors, Micro-Iris
Curved, sharp/sharp, very delicate, 90 mm
Catalog # HAMMHSB545-09
Supplier:   HAMMACHER KARL
Iris scissors, Micro-Iris
Catalog # HAMMHSB545-09
Supplier:  HAMMACHER KARL
Iris scissors, Micro-Iris
Iris scissors, Micro-Iris
Curved, sharp/sharp, very delicate, 90 mm
Price
71,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
These iris scissors are made from stainless steel.
Specifications
  • Type:
    Curved, sharp/sharp, very delicate
  • Length:
    90 mm
  • Seq:
    02