Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Micro scissors, model Tübingen

Tübingen, curved, sharp/sharp, cutting length 7 mm, 85 mm
Catalog # HAMMHSB531-08
Supplier:   HAMMACHER KARL
Micro scissors, model Tübingen
Catalog # HAMMHSB531-08
Supplier:  HAMMACHER KARL
Micro scissors, model Tübingen
Micro scissors, model Tübingen

Tübingen, curved, sharp/sharp, cutting length 7 mm, 85 mm

Price
472,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel.

Specifications
  • Type:
    Tübingen, curved, sharp/sharp, cutting length 7 mm
  • Length:
    85 mm
  • Seq:
    05