Orders LinkContactUsLinkComponent
Micro scissors
Straight, sharp/sharp, 90 mm
  HAMMHSB516-09
Micro scissors
  HAMMHSB516-09
Micro scissors
Micro scissors
Straight, sharp/sharp, 90 mm
384,00 €
1 * 1 ST
Stainless steel.
  • Type:
    Straight, sharp/sharp
  • Length:
    90 mm
  • Seq:
    07