Order Entry
Austria
Orders LinkContactUsLinkComponent
Electrolyte Solutions

Electrolyte Solutions

Electrolyte KCl 3 M, 100 ml, 100 ml
Catalog # 662-9202
Supplier:   HAMILTON BONADUZ
Electrolyte Solutions
Catalog # 662-9202
Supplier:  HAMILTON BONADUZ
CAS Number:  
Electrolyte Solutions
Electrolyte Solutions

Electrolyte KCl 3 M, 100 ml, 100 ml

CAS Number:  
Price
15,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

A variety of electrolyte solutions for pH and ORP electrodes.

Documents

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    Electrolyte KCl 3 M, 100 ml

Specifications