Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Pestles

55 mm, 210 mm, 410-9124
  410-9135
Pestles
  410-9135
Pestles
Pestles

55 mm, 210 mm, 410-9124

 
51,80 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9124
  • Head Ø:
    55 mm
  • Length:
    210 mm
  • Seq:
    19