Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Pestles

42 mm, 175 mm, 410-9112
  410-9134
Pestles
  410-9134
Pestles
Pestles

42 mm, 175 mm, 410-9112

 
39,30 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9112
  • Head Ø:
    42 mm
  • Length:
    175 mm
  • Seq:
    16