Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Pestles

36 mm, 150 mm, 410-9110
  410-9133
Pestles
  410-9133
Pestles
Pestles

36 mm, 150 mm, 410-9110

 
27,40 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9110
  • Head Ø:
    36 mm
  • Length:
    150 mm
  • Seq:
    15