Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Pestles

24 mm, 115 mm, 410-9326
  410-9131
Pestles
  410-9131
Pestles
Pestles

24 mm, 115 mm, 410-9326

 
15,60 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9326
  • Head Ø:
    24 mm
  • Length:
    115 mm
  • Seq:
    12