Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Büchner funnels

4300 ml, 240 mm
Catalog # 511-2511
Büchner funnels
Catalog # 511-2511
Büchner funnels
Büchner funnels

4300 ml, 240 mm

Price
649,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

White glazed porcelain.

For filter paper or membrane filtration discs.

 : DIN 12 905 (511-2502, 511-2501 and 511-2500 not according to DIN).

Specifications
  • Capacity:
    4300 ml
  • Seq:
    12
  • For filter Ø:
    240 mm