Order Entry
Austria
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

50 ml, 45 mm
Catalog # 511-2503
Büchner funnels
Catalog # 511-2503
Büchner funnels
Büchner funnels

50 ml, 45 mm

Price
56,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

White glazed porcelain.

For filter paper or membrane filtration discs.

 : DIN 12 905 (511-2502, 511-2501 and 511-2500 not according to DIN).

Specifications
  • Capacity:
    50 ml
  • Seq:
    04
  • For filter Ø:
    45 mm