Order Entry
Austria
Orders LinkContactUsLinkComponent
Cell scrapers

Cell scrapers

Cell scraper with 18 mm pivoted blade, 280 mm
Catalog # 391-3010
Supplier:   Greiner Bio-One
Cell scrapers
Catalog # 391-3010
Supplier:  Greiner Bio-One
Cell scrapers
Cell scrapers

Cell scraper with 18 mm pivoted blade, 280 mm

Price
169,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

Designed to facilitate the gentle mechanical removal of adherent cells from growth surfaces. Each scraper has a 18 mm pivoted (pivot angle 60°) scraper blade, which allows access to poorly accessible surfaces.

  • Optimised blade design for maximum cell harvest
  • Uniform contact with the growth surface, minimal mechanical strain and efficient cell harvest, even from poorly accessible surfaces
  • Minimises any accumulation of the cell suspension to the blade structure

Packaging: Individually packaged.

Documents

Specifications
  • Description:
    Cell scraper with 18 mm pivoted blade
  • Total length:
    280 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.