Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Buffer solutions

Buffer solution, 8,00, ±0,01, 20 °C, Plastic bottle, 5 L
  REGE5080
 :   REAGECON
Buffer solutions
  REGE5080
 :  REAGECON
 :  
Buffer solutions
Buffer solutions

Buffer solution, 8,00, ±0,01, 20 °C, Plastic bottle, 5 L

 :  
 
141,00 €
   
1 * 5 L
 

 

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    8,00