Buffer solutions
Buffer solution, 7,00, ±0,01, 20 °C, Plastic bottle, 5 L
Catalog # REGE5070
Supplier: REAGECON
Jump to:
Product Details & Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Packung:Plastic bottle
- VE:5 L
- Referenztemperatur:20 °C
- Bezeichnung:Buffer solution
- pH Genauigkeit:±0,01
- pH-Wert:7,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



