Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solution, 4,00, ±0,01, 20 °C, Plastic bottle, 5 L
Catalog # REGE5040
Supplier:   REAGECON
Buffer solutions
Catalog # REGE5040
Supplier:  REAGECON
CAS Number:  
Buffer solutions
Buffer solutions

Buffer solution, 4,00, ±0,01, 20 °C, Plastic bottle, 5 L

CAS Number:  
Price
127,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,01
  • pH value:
    4,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.