BUFFER SOLUTION PH 6.881 @ 20C NIST 1 * 500 mL
Catalog # REGE106881
Supplier: REAGECON
Frequently Bought Together
Cleanroom wipes, pre-saturated, SATWIPES® Theta
Supplier: CONTEC
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH electrode storage and cleaning solutions, Orion™
Supplier: Thermo Orion
VWR®, Nitrile Gloves
Supplier: VWR Collection
Calcium carbonate, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Laboratory trays
Supplier: KARTELL
L(+)-Ascorbic acid 99.0-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach