BUFFER PH 4 ROOD 20-25 C 1 * 500 mL
Catalog # REGE10405CTT
Supplier: REAGECON
Frequently Bought Together
BUFFER PH 7 GEEL 20-25 C 1 * 500 mL
Supplier: REAGECON
Labelling tapes, temperature resistant
Supplier: PRECISION DYNAMICS TIMEM
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Measuring Cylinders, Class A, TPX
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR®, Electrode cleaning solutions
Supplier: VWR Chemicals
BUFFER SOLUTION PH10 TWIN NECK 1 * 500 mL
Supplier: REAGECON
Sodium dodecyl sulphate (SDS) 99% for biochemistry
Supplier: Thermo Fisher Scientific