ContactUsLinkComponent
Endocrine System. - 5 Microscope Slides - 1. Pituitary gland (hypophysis), human, t.s. 2. Pineal gland (epiphysis), sheep, l.s. 3. T hyroid gland, sheep, t.s. 4. Pancreas with islets of Langerhans, cat, t.s. 5. Adrenal gland, cat, t.s. 6. Corpus lute 1 * 6 items
Catalog # LIED70600
Supplier:  LIEDER JOHANNES
Endocrine System. - 5 Microscope Slides - 1. Pituitary gland (hypophysis), human, t.s. 2. Pineal gland (epiphysis), sheep, l.s. 3. T hyroid gland, sheep, t.s. 4. Pancreas with islets of Langerhans, cat, t.s. 5. Adrenal gland, cat, t.s. 6. Corpus lute 1 * 6 items
Catalog # LIED70600
Supplier:  LIEDER JOHANNES
Frequently Bought Together