Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 15, excitation BP 546/12, beam splitter FT 580, emission LP 590
Catalog # 630-1422
Supplier:   ZEISS
Filter sets
Catalog # 630-1422
Supplier:  ZEISS
Filter sets
Filter set 15, excitation BP 546/12, beam splitter FT 580, emission LP 590
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Filter set 15, excitation BP 546/12, beam splitter FT 580, emission LP 590
  • Seq:
    03