Order Entry
Austria
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Long life halogen bulb, 6 V, 20 W
Catalog # 630-0322
Supplier:   ZEISS
Halogen bulbs
Catalog # 630-0322
Supplier:  ZEISS
Halogen bulbs

Long life halogen bulb, 6 V, 20 W

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Long life halogen bulb
  • Seq:
    07
  • Power:
    20 W
  • Voltage:
    6 V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.