Order Entry
Austria
Orders LinkContactUsLinkComponent
Halogen bulbs

Halogen bulbs

Halogen bulb, 6 V, 30 W
Catalog # 630-1711
Supplier:   ZEISS
Halogen bulbs
Catalog # 630-1711
Supplier:  ZEISS
Halogen bulbs

Halogen bulb, 6 V, 30 W

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Halogen bulb
  • Seq:
    01
  • Power:
    30 W
  • Voltage:
    6 V