ContactUsLinkComponent
Tetracon® 700-15,universal 4 electrode conductivity cell especially for waste water treatment plants, for use with ecoline and quadr oline® monitors with integrated temperature sensor, 15 m cable and watertight plug (ip68) 1 * 1 items
Catalog # WTWO302318
Supplier:  WTW ONLINE
Tetracon® 700-15,universal 4 electrode conductivity cell especially for waste water treatment plants, for use with ecoline and quadr oline® monitors with integrated temperature sensor, 15 m cable and watertight plug (ip68) 1 * 1 items
Catalog # WTWO302318
Supplier:  WTW ONLINE
Frequently Bought Together