Orders LinkContactUsLinkComponent
External Printer
External Printer
Serial port RS232, for external printer
Catalog # SMEGWD-232PRINT
New Product
Supplier:   SMEG
External Printer
Catalog # SMEGWD-232PRINT
Supplier:  SMEG
New Product
External Printer
Serial port RS232, for external printer
Price
126,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Externla printer includes RS232 serial port card.
  • Dot matrix thermal printing
    Complete with clock function
    Equipped with independent power supply
    For use with GW4260 series
To be combined with a roll of thermal paper Ø 50 mm and width 57 mm.
Specifications
  • Description:
    Serial port RS232, for external printer
  • Seq:
    2