Orders LinkContactUsLinkComponent
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
Technicloth® floor mop wipers, 690×380 mm
Catalog # 115-0480
New Product
Supplier:   Texwipe
TechniCloth® Flat Mop Covers
Catalog # 115-0480
Supplier:  Texwipe
New Product
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
Technicloth® floor mop wipers, 690×380 mm
Price
462,06 €
Log in for availability
1 * 4 ST
Product Details & Documents
Mop covers (ISO Class 3 - 8) for cleaning and decontaminating floors.
  • Made without chemical binders or additives
  • Non-woven, economical polyester/cellulose for cleaning and removing residues
  • Chemically resistant, withstand exposure to common cleaning and disinfecting solutions
Specifications
  • Description:
    Technicloth® floor mop wipers
  • L×W:
    690×380 mm
  • Seq:
    5